Anti-GRIA3 Rabbit Polyclonal Antibody
Catalog # AVIVARP35009_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor 3
- Antigen symbol:GRIA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:29628
- Antigen synonyms:GluR-3|GluA3|GLUR3|GluR-K3|GluR-C
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GRIA3 (glutamate receptor, ionotropic, AMPA 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat