Anti-CACNB3 Rabbit Polyclonal Antibody
AVIVARP34957_T100
- Antibody type:Primary
- Antigen name:calcium channel, voltage-dependent, beta 3 subunit
- Antigen symbol:CACNB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:784
- Antigen synonyms:CACNLB3|CAB3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: CACNB3 (calcium channel, voltage-dependent, beta 3 subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: