Anti-ZNF568 Rabbit Polyclonal Antibody
AVIVARP34877_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 568
- Antigen symbol:ZNF568
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:374900
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF568 (zinc finger protein 568)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: