Anti-WDR45B Rabbit Polyclonal Antibody
Catalog # AVIVARP34827_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:WD repeat domain 45B
- Antigen symbol:WDR45B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:56270
- Antigen synonyms:WIPI3|WIPI-3|WDR45L
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: WDR45B (WD repeat domain 45B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: