Anti-CTRB1 Rabbit Polyclonal Antibody
Catalog # AVIVARP34022_P050
Supplier: Aviva Systems Biology
Jump to:
Product Details & Documents
Type: Primary
Antigen: CTRB1 (chymotrypsinogen B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Documents
ARP34022_P050
Datasheet
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Epitope:middle
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:Chymotrypsinogen B1
- Cat. No.:AVIVARP34022_P050
- Reactivity:Human|Bovine|Rat|Mouse
- Gene ID:1504
- Antigen synonyms:CTRB
- Conjugation:Unconjugated
- Antigen symbol:CTRB1



