Anti-FABP3 Rabbit Polyclonal Antibody
Catalog # AVIVARP33823_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 3, muscle and heart
- Antigen symbol:FABP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2170
- Antigen synonyms:O-FABP|FABP11|MDGI|H-FABP|M-FABP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: FABP3 (fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human