Anti-C4BPB Rabbit Polyclonal Antibody
Catalog # AVIVARP33814_P050
Supplier: Aviva Systems Biology
Jump to:
Product Details & Documents
Type: Primary
Antigen: C4BPB (complement component 4 binding protein, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Epitope:N-terminal
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:complement component 4 binding protein, beta
- Cat. No.:AVIVARP33814_P050
- Gene ID:725
- Antigen synonyms:C4BP
- Conjugation:Unconjugated
- Antigen symbol:C4BPB



