Anti-EPB42 Rabbit Polyclonal Antibody
Catalog # AVIVARP33799_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:erythrocyte membrane protein band 4.2
- Antigen symbol:EPB42
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:2038
- Antigen synonyms:PA|SPH5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EPB42 (erythrocyte membrane protein band 4.2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: