Anti-NR0B1 Rabbit Polyclonal Antibody
AVIVARP32871_P050
- Antibody type:Primary
- Antigen name:Nuclear receptor subfamily 0, group B, member 1
- Antigen symbol:NR0B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:11614
- Antigen synonyms:Ahch|DAX-1|Ahc|AHX|Dax1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NR0B1 (nuclear receptor subfamily 0, group B, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: