Anti-DBI Rabbit Polyclonal Antibody
AVIVARP32803_P050
- Antibody type:Primary
- Antigen name:diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein)
- Antigen symbol:DBI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:1622
- Antigen synonyms:EP|CCK-RP|ACBD1|ACBP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DBI (diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human