Anti-IKZF1 Rabbit Polyclonal Antibody
Catalog # AVIVARP32758_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:IKAROS family zinc finger 1 (Ikaros)
- Antigen symbol:IKZF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:10320
- Antigen synonyms:PRO0758|ZNFN1A1|LyF-1|PPP1R92|LYF1|IK1|IKAROS|Hs.54452
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: IKZF1 (IKAROS family zinc finger 1 (Ikaros))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: