Anti-AKAP8 Rabbit Polyclonal Antibody
AVIVARP32754_P050
- Antibody type:Primary
- Antigen name:A kinase (PRKA) anchor protein 8
- Antigen symbol:AKAP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:10270
- Antigen synonyms:AKAP95|AKAP-8|AKAP-95|AKAP 95
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: AKAP8 (A kinase (PRKA) anchor protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: