Anti-TRIM23 Rabbit Polyclonal Antibody
Catalog # AVIVARP32672_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 23
- Antigen symbol:TRIM23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:373
- Antigen synonyms:RNF46|ARFD1|ARD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TRIM23 (tripartite motif containing 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: