Anti-APEX1 Rabbit Polyclonal Antibody
Catalog # AVIVARP32651_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:APEX nuclease (multifunctional DNA repair enzyme) 1
- Antigen symbol:APEX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:328
- Antigen synonyms:APEX|APEN|REF1|APE|APE1|APX|HAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: