Anti-AIRE Rabbit Polyclonal Antibody
AVIVARP32650_P050
- Antibody type:Primary
- Antigen name:autoimmune regulator
- Antigen symbol:AIRE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:326
- Antigen synonyms:AIRE1|APECED|PGA1|APSI|APS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: