Anti-CHEK1 Rabbit Polyclonal Antibody
Catalog # AVIVARP32589_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Checkpoint Kinase 1
- Antigen symbol:CHEK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1111
- Antigen synonyms:CHK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: