Anti-SND1 Rabbit Polyclonal Antibody
Catalog # AVIVARP32563_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Staphylococcal nuclease and tudor domain containing 1
- Antigen symbol:SND1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:27044
- Antigen synonyms:p100|TDRD11
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SND1 (staphylococcal nuclease and tudor domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: