Anti-AKAP8L Rabbit Polyclonal Antibody
Catalog # AVIVARP32560_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:A kinase (PRKA) anchor protein 8-like
- Antigen symbol:AKAP8L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:26993
- Antigen synonyms:HAP95|HA95|NAKAP95|NAKAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: AKAP8L (A kinase (PRKA) anchor protein 8-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: