Anti-GBX2 Rabbit Polyclonal Antibody
Catalog # AVIVARP32552_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:gastrulation brain homeobox 2
- Antigen symbol:GBX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:2637
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GBX2 (gastrulation brain homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: