Anti-SPDEF Rabbit Polyclonal Antibody
Catalog # AVIVARP32534_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:SAM pointed domain containing ETS transcription factor
- Antigen symbol:SPDEF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:25803
- Antigen synonyms:PDEF|bA375E1.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SPDEF (SAM pointed domain containing ETS transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: