Order Entry
Export
ContactUsLinkComponent
Anti-FOSB Rabbit Polyclonal Antibody
Anti-FOSB Rabbit Polyclonal Antibody
  AVIVARP32519_T100
undefined
Anti-FOSB Rabbit Polyclonal Antibody
  AVIVARP32519_T100

 

  • Antibody type:
    Primary
  • Antigen name:
    FBJ murine osteosarcoma viral oncogene homolog B
  • Antigen symbol:
    FOSB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    2354
  • Antigen synonyms:
    AP-1|GOSB|G0S3|GOS3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: FOSB (FBJ murine osteosarcoma viral oncogene homolog B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: