Anti-SEC14L2 Rabbit Polyclonal Antibody
Catalog # AVIVARP32518_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:SEC14-like lipid binding 2
- Antigen symbol:SEC14L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23541
- Antigen synonyms:C22orf6|TAP1|SPF|TAP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SEC14L2 (SEC14-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: