Anti-EHMT2 Rabbit Polyclonal Antibody
Catalog # AVIVARP32493_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Euchromatic histone-lysine N-methyltransferase 2
- Antigen symbol:EHMT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10919
- Antigen synonyms:GAT8|NG36|C6orf30|BAT8|KMT1C|G9A
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: EHMT2 (euchromatic histone-lysine N-methyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: