Anti-POU3F2 Rabbit Polyclonal Antibody
AVIVARP31218_P050
- Antibody type:Primary
- Antigen name:POU class 3 homeobox 2
- Antigen symbol:POU3F2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:5454
- Antigen synonyms:BRN2|OTF-7|OTF7|POUF3|oct-7|brn-2|OCT7|N-Oct3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: POU3F2 (POU class 3 homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: