Anti-E2F1 Rabbit Polyclonal Antibody
Catalog # AVIVARP31171_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:E2F transcription factor 1
- Antigen symbol:E2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:1869
- Antigen synonyms:RBP3|RBAP1|E2F-1|RBBP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: E2F1 (E2F transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human