Anti-ARRB1 Rabbit Polyclonal Antibody
Catalog # AVIVARP30523_P050
Supplier: Aviva Systems Biology
Jump to:
Product Details & Documents
Type: Primary
Antigen: ARRB1 (arrestin, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Documents
ARP30523_P050.pdf
Datasheet
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Epitope:middle
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:Arrestin beta 1
- Cat. No.:AVIVARP30523_P050
- Gene ID:408
- Antigen synonyms:ARB1|ARR1
- Conjugation:Unconjugated
- Antigen symbol:ARRB1



