Anti-CD163L1 Rabbit Polyclonal Antibody
BOSSBS-7993R
: Bioss
- Antibody type:Primary
- Antigen name:CD163 molecule-like 1
- Antigen symbol:CD163L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CD163B|M160
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: CD163L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human