Anti-HSD11B1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3904R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid 11-beta Dehydrogenase 1
- Antigen symbol:HSD11B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:corticosteroid 11-beta-dehydrogenase isozyme 1|HSD11B1|11-beta-hydroxysteroid dehydrogenase 1|HSD11L|HDL|MGC13539|short chain dehydrogenase/reductase family 26C|OTTHUMP00000034650|hydroxysteroid (11-beta) dehydrogenase 1|SDR26C1|HSD11B|member 1|11-DH|HSD11|11-beta-HSD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: HSD11B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat