Anti-SIRT1 Rabbit Polyclonal Antibody
BOSSBS-3393R
: Bioss
- Antibody type:Primary
- Antigen name:Sirtuin 1
- Antigen symbol:SIRT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:NAD-dependent Deacetylase Sirtuin-1|SIR2L1|SIRT1|SIR2-like Protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: SIRT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse