Anti-CLDN4 Rabbit Polyclonal Antibody
BOSSBS-2790R
: Bioss
- Antibody type:Primary
- Antigen name:Claudin 4
- Antigen symbol:CLDN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:WBSCR8|CPE-R|hCPE-R|CPETR|CPETR1|CPER
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: CLDN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: