Anti-PSMB9 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2787R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 20S Beta 9
- Antigen symbol:PSMB9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:macropain) subunit|proteasome-related gene 2|low molecular mass protein 2|9 (large multifunctional peptidase 2)|beta type|macropain chain 7|proteasome catalytic subunit 1i|proteasome beta 9 subunit|proteasome (prosome|RING12|MGC70470|LMP2|proteasome chain 7|beta1i|PSMB6i|proteasome subunit beta 6i|multicatalytic endopeptidase complex chain 7|PSMB9
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PSMB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat