Anti-IL1B Rabbit Polyclonal Antibody
BOSSBS-0812R
: Bioss
- Antibody type:Primary
- Antigen name:Interleukin 1 beta
- Antigen symbol:IL1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:catabolin|IL-1 beta|preinterleukin 1 beta|IL1 B|pro-interleukin-1-beta|IL1-BETA|interleukin-1 beta|IL-1|IL1B|IL1F2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog