Anti-TNNI3 Rabbit Polyclonal Antibody
BOSSBS-0799R
: Bioss
- Antibody type:Primary
- Antigen name:Troponin I Type 3
- Antigen symbol:TNNI3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:CMD2A|RCM1|TNNC1|CMD1FF|cTnI|CMH7
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: TNNI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat