Anti-KCNMA1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-0729R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Potassium Large Conductance Calcium-activated Channel Subfamily M Alpha member 1
- Antigen symbol:KCNMA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:none
- Antigen synonyms:BKTM|mSLO1|MaxiK|SLO|SLO-ALPHA|KCa1.1|bA205K10.1|SAKCA|SLO1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: KCNMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat