ContactUsLinkComponent
Industrial gloves, Camapur® Comfort 619
Industrial gloves, Camapur® Comfort 619
Supplier:  KCL by Honeywell
Certificates
undefined
Industrial gloves, Camapur® Comfort 619
Supplier:  KCL by Honeywell
Specifications
  • Catalog No:
  • KCHL619.6
  • KCHL619.7
  • KCHL619.10
  • KCHL619.8
  • KCHL619.9
  • Model:
  • Comfort 619
  • Comfort 619
  • Comfort 619
  • Comfort 619
  • Comfort 619
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
  • EN 388:2016 (3131X)
Specifications