Order Entry
Export
ContactUsLinkComponent
Sterilisation indicators
Sterilisation indicators
  489-0013
 :  PHAGOGENE
Sterilisation indicators
  489-0013
 :  PHAGOGENE

 

  • Description:
    Sterilisation indicator, TST 3702 "standard cycle": 125 °C for 20 minutes or 134 °C for 9 minutes
  • Seq:
    02
  • Pk:
    200

 

 

The indicator turns from yellow to blue upon autoclaving.

  • Non-sequential indicator
  • Non-toxic organic ink
  • Declination according to cycles of autoclaving: 'short cycle', 'standard cycle' or 'prion cycle'
 : Compliant with ISO standards.