Diaphragm vacuum pumps, DIVAC L
Supplier: LEYBOLD
Certificates
About this item
DIVAC L two stage diaphragm pumps are designed for almost all requirements in the chemistry lab. Pumps are basically corrosion and solvent resistant since their parts in contact with the pumped medium are made of PTFE (Teflon), FFPM (Kalrez) and PVDF (Solef).
Delivery: Supplied with 2,3 meter cable and Schuko plug.