Orders LinkContactUsLinkComponent
Sulphate test kit, SF-1
Sulphate test kit, SF-1
Sulphate test kit, Model SF-1, 50 - 200 mg/l, 1 KIT
Catalog # HACH225100
Supplier:   Hach
Sulphate test kit, SF-1
Catalog # HACH225100
Supplier:  Hach
CAS Number:  
Sulphate test kit, SF-1
Sulphate test kit, SF-1
Sulphate test kit, Model SF-1, 50 - 200 mg/l, 1 KIT
CAS Number:  
Price
214,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
High concentrations of magnesium or sodium sulphate in water act as cathartics. This test kit provides economical determination of sulphate levels using the Extinction/Turbidimetric methods.
Delivery information: Includes graduated cylinder, mixing bottle, reagent, dipstick comparator, instructions and carrying case.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Sulphate test kit, Model SF-1
  • Range:
    50 - 200 mg/l
Specifications