ContactUsLinkComponent
Agarose Sealing Solution
Agarose Sealing Solution
Catalog # 786-226
Supplier:  G-Biosciences
undefined
Agarose Sealing Solution
Catalog # 786-226
Supplier:  G-Biosciences
Specifications
  • Description:
    Agarose Sealing Solution
  • Pk:
    50 mL
Specifications
About this item
Simply heat and use for sealing the IPG strips for SDS-PAGE analysis. The Agarose Sealing Solution is prepared in a proprietary buffer to minimise reoxidation of the competing thiol pairs as proteins enter into the second dimension gel. Improves resolution and prevents streaking of protein spots on 2D gels.