Diethyl pyrocarbonate
Supplier: G-Biosciences
Jump to:
About this item
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
- CAS Number:1609-47-8
- Formula:O(COOC₂H₅)₂
- MDL Number:MFCD00009106
- Molecular Weight:162.14 g/mol
- Merck Index:13,08088
- Boiling Pt:93 °C (1013 hPa)
- Density:1.123 g/cm³ (20 °C)
- Flash Pt:69 °C
- Storage Temperature:Refrigerator
- EINECS:216-542-8
2 Options Available Below
Product Details & Documents
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
- CAS Number:1609-47-8
- Formula:O(COOC₂H₅)₂
- MDL Number:MFCD00009106
- Molecular Weight:162.14 g/mol
- Merck Index:13,08088
- Boiling Pt:93 °C (1013 hPa)
- Density:1.123 g/cm³ (20 °C)
- Flash Pt:69 °C
- Storage Temperature:Refrigerator
- EINECS:216-542-8



