ContactUsLinkComponent
Tubing, vacuum
Tubing, vacuum
Catalog # LEYB86803
Supplier:  LEYBOLD
Tubing, vacuum
Catalog # LEYB86803
Supplier:  LEYBOLD
Specifications
  • Description:
    With flanges DN 25 ISO-KF, 1000 mm
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
Stainless steel 1.4571.
  • Excellent chemical resistance
  • Excellent temperature resistance
With flange connection