Platinum wire
Supplier: CHEMPUR
Jump to:
About this item
Platinum 99,9%.
Platinum wire is used in many applications including gasket or O-ring fabrication, thermocouples, electrical contacts, loops and heater coils.
5 Options Available Below
Product Details & Documents
Platinum 99,9%.
Platinum wire is used in many applications including gasket or O-ring fabrication, thermocouples, electrical contacts, loops and heater coils.