Order Entry
Export
ContactUsLinkComponent
undefined | Protein Purification Resins
Catalog # 555-0066
CAS Number:  
undefined
Catalog # 555-0066
CAS Number:  

Specifications

  • Storage temperature:
    2 - 8 °C
  • Exclusion limit:
    >5000000 Da
  • Cat. no.:
    555-0066

Specifications

About this item