Diaphragm vacuum gauge DIAVAC DV 1000
Catalog # 188-1501
Supplier: LEYBOLD
Specifications
- Measurement range:1 - 1000 mbar
- Description:Diaphragm vacuum gauge, DIAVAC DV 1000, with mechanical display
- Model:DV 1000
- Permitted ambient temperature for operation:0...60 ° C
- Permitted ambient temperature for storage:−10...60 °C
- Vacuum connection:DN 40 ISO-KF
- Weight:2,7 kg
- W×D×H:180×166×100 mm
- Pk:1
Specifications
About this item
Rugged mechanical diaphragm vacuum gauge of high accuracy for the rough vacuum range from 1 to 1000 mbar (1 to 750 Torr).
Typical applications: chemical processes, vacuum destillation, absolute pressure measurements, for gas mixtures, for use in explosion hazard rated areas, drying processes, lamp manufacture.
- Wide measurement range
- The scale of each gauge is individually calibrated
- Absolute pressure gauge
- Readout independent of the type of gas and changes in atmospheric pressure
- Stainless steel diaphragm for excellent compatibility with most media
- Laser welding technology for high precision diaphragm mount
- Rugged table-top housing, can be freely mounted above the flange connection; also for panel mounting
- Measurement chamber can be easily cleaned owing to the detachable measurement flange