ContactUsLinkComponent
Scrubbing brush
Scrubbing brush
Catalog # NEOL2-3275
Supplier:  NEOLAB
undefined
Scrubbing brush
Catalog # NEOL2-3275
Supplier:  NEOLAB
Specifications
  • Handle material:
    Wood
  • Description:
    Scrubbing brush, without handle
  • Total length:
    220 mm
  • Pk:
    1
  • Trim length:
    50 mm
Specifications
About this item
Scrubbing brush with fibre bristles and wooden block.