Kofferdam dental punch, Ainsworth, Aesculap®
Catalog # AESCDF401R
Supplier: Aesculap
Specifications
- Description:Kofferdam dental punch, Ainsworth
- Length:160 mm
- Pk:1
Specifications
About this item
Reusable, non sterile dental punch for Maxillo-facial surgery and dental surgery.