ContactUsLinkComponent
Ear punches, IMS
Ear punches, IMS
Catalog # HARV34-0136
Supplier:  HARVARD APPARATUS
Ear punches, IMS
Catalog # HARV34-0136
Supplier:  HARVARD APPARATUS
Specifications
  • Ø:
    1,5 mm
  • Description:
    Forceps type punch, model 4.7.1
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This easily operated hole punching device is suitable for marking small rodents whenever a permanent identification system is required. Two types of punches are available. A forceps type which provides a longer handle for gripping, or the simple thumb type.
Packaging: These punches are supplied individually.