Wound plasters, Aluderm®-aluplast waterproof
Supplier: SOEHNGEN
Certificates
About this item
The aluderm®-aluplast wound plaster has an aluminium wound layer and an additional absorption middle layer. The aluminised wound layer is highly absorbent, so the wound remains dry and clean.
- Made from PE plastic with polyacryl adhesive
- Adhesive function is activated by body heat
- Plaster is transparent, durable and waterproof
- Recommended in places where water and dirt are present
- Individually sealed