Cable ties, releasable, Allplastik-Blitzbinder®
Supplier: WUERTEMBERGISCHE ALLPLASTIK
Certificates
About this item
The Allplastik-Blitzbinder® quick fasteners are made of PE and have a special dual closure design.
- Provides maximum strength and reliable closing when combined with a round profile
- Can be opened again at any time
- Allows a hanging loop to be formed
- Temperature resistance: −40 to +70 °C
Ideal for use as sack fastener, bag fastener, releasable cable tie, plant tie, general-purpose binder, quick fastener, fixing tie, bag tie, scaffolding tie, tarpaulin tie, cable tie for bundling cable harnesses.
Certifications: FDA compliant