Glove liners, Puri Palm Fit
: PURITECH
These glove liners made of 100% soft nylon fabric with PU coating on the palm are ideal for handling small components.
- Seamless liner
- Hand-specific for great fit
- Excellent dexterity and high absorbency
Can be used for light dry assembly, maintenance or micro mechanical tasks and for pharmaceutical processing.
Thickness of coating: 0,1 mm.
- Catalog No:
- 112-4187
- 112-4184
- 112-4183
- 112-4182
- 112-4185
- 112-4186
- Model:
- Palm Fit
- Palm Fit
- Palm Fit
- Palm Fit
- Palm Fit
- Palm Fit
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist